MX
ES
Aplicaciones
Productos
Servicios
Asistencia técnica
Home
Search Results
Mechanical and structural contribution of non-fibrillar matrix in uniaxial tension: a collagen-agarose co-gel model
Mechanical and structural contribution of non-fibrillar matrix in uniaxial tension: a collagen-agarose co-gel model
Annals of Biomedical Engineering (2011)
Lake SP and Barocas VH