New Insights into the Cystine-Sulfite Reaction.
Molecules (Basel, Switzerland) (2019-06-30)
Matteo Zecchini, Robert Lucas, Adam Le Gresley
RESUMEN
The mechanism by which cysteine-S-sulfate is formed from the reaction of sulfite with cystine in the absence of a dedicated oxidizing agent is investigated using high-resolution NMR. Changes to reactant ratio, pH, UV light exposure and temperature were evaluated to determine the most effective conditions to achieve the maximum yield of cysteine-S-sulfate without recourse to conventional oxidizing reagents. Herein evidence is provided for both nucleophilic and radical mechanisms, by which cysteine-S-sulfate can be generated with yields of up to 96%.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)