  • Search Results
  • Validation of an off line solid phase extraction liquid chromatography?tandem mass spectrometry method for the determination of systemic insecticide residues in honey and pollen samples collected in apiaries from NW Spain.
Validation of an off line solid phase extraction liquid chromatography?tandem mass spectrometry method for the determination of systemic insecticide residues in honey and pollen samples collected in apiaries from NW Spain.
Analytica Chimica Acta (2010)
Garcia-Chao M, et al.