Activation of Raf-1 signaling by protein kinase C through a mechanism involving Raf kinase inhibitory protein.
The Journal of biological chemistry (2003-01-29)
Kevin C Corbit, Nicholas Trakul, Eva M Eves, Bruce Diaz, Mark Marshall, Marsha Rich Rosner
RESUMEN
Protein kinase C (PKC) regulates activation of the Raf-1 signaling cascade by growth factors, but the mechanism by which this occurs has not been elucidated. Here we report that one mechanism involves dissociation of Raf kinase inhibitory protein (RKIP) from Raf-1. Classic and atypical but not novel PKC isoforms phosphorylate RKIP at serine 153 (Ser-153). RKIP Ser-153 phosphorylation by PKC either in vitro or in response to 12-O-tetradecanoylphorbol-13-acetate or epidermal growth factor causes release of RKIP from Raf-1, whereas mutant RKIP (S153V or S153E) remains bound. Increased expression of PKC can rescue inhibition of the mitogen-activated protein (MAP) kinase signaling cascade by wild-type but not mutant S153V RKIP. Taken together, these results constitute the first model showing how phosphorylation by PKC relieves a key inhibitor of the Raf/MAP kinase signaling cascade and may represent a general mechanism for the regulation of MAP kinase pathways.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-HA−Peroxidase antibody, Mouse monoclonal antibody produced in mouse, clone HA-7, purified from hybridoma cell culture