Axotomy-induced cytoskeleton changes in unmyelinated mammalian central nervous system axons.
Neuroscience (2011-01-11)
C Balaratnasingam, W H Morgan, L Bass, M Kang, S J Cringle, D-Y Yu
RESUMEN
Oligodendrocyte-derived myelin retards the ability of CNS axons to regenerate following transection. The intrinsic response of CNS axons to an axotomy insult may be vastly different in the absence of myelin. However, the paucity of adequate experimental models has limited detailed investigation of cellular behaviour following axon transection in an unmyelinated CNS environment. In this study we perform laser-induced axotomy of the porcine retinal ganglion cell axon, a physiologically unmyelinated, mature CNS axon that is structurally similar to humans to infer knowledge about axonal behaviour in the absence of myelin. Axotomy-induced changes to the neuronal cytoskeleton and supporting astrocytes during the early stages after transection are delineated by examining the sequence of neurofilament subunit, microtubule (TUB), microtubule associated protein (MAP), glial fibrillary acidic protein (GFAP) and terminal deoxynucleotidyl transferase biotin-dUTP nick end labelling (TUNEL) modification. Axonal transection induced an increase in the expression of neurofilament light at regions within, and immediately adjacent to, sites of axotomy. Other neurofilament subunits were not altered at sites of transection. Unlike myelinated axons where an increase in GFAP staining within hypertrophic glial scars have been shown to inhibit axonal repair we demonstrate a decrease in GFAP staining within regions of increased or preserved neurofilament expression. The behaviour of TUB and MAP proteins following transection of unmyelinated CNS axons are similar to what has previously been described in myelinated CNS axons. This study provides fundamental insights into astrocyte and axonal behaviour acutely after axotomy and demonstrates a series of degenerative events in unmyelinated CNS axons, which in comparison to prior reports are different to myelinated CNS axons. The findings of this report have relevance to understanding pathogenic mechanisms underlying neuro-degeneration in the CNS.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 (Phos. and Non-Phos.) antibody produced in mouse, clone N52, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 antibody produced in mouse, clone NE14, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Neurofilament 160 antibody produced in mouse, clone NN18, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Tubulin I+II antibody produced in mouse, clone JDR.3B8, ascites fluid
Sigma-Aldrich
Monoclonal Anti-MAP1 antibody produced in mouse, clone HM-1, ascites fluid
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.