YY1 cooperates with TFEB to regulate autophagy and lysosomal biogenesis in melanoma.
Molecular carcinogenesis (2019-08-27)
Jing Du, Wenyan Ren, Fengping Yao, Hong Wang, Kexin Zhang, Meiying Luo, Yuxue Shang, Douglas O'Connell, Zhuchun Bei, Hongquan Wang, Ran Xiong, Yongfei Yang
RESUMEN
Autophagy is a self-proteolytic process that degrades intracellular material to maintain cellular homeostasis. Transcription factor EB (TFEB) is the master activator that regulates the transcription of genes involved in autophagy and lysosomal biogenesis. However, the cotranscriptional factors of TFEB are rarely identified. Here, we found that Yin Yang 1 (YY1) regulated autophagy and lysosome biogenesis in melanoma cells. YY1 cooperates with TFEB to regulate autophagy through controlling the transcription of autophagy and lysosome biogenesis related genes. Moreover, suppression of YY1 enhanced the antitumor efficiency of vemurafenib both in vitro and in vivo. Collectively, these studies identify YY1 as a novel cotranscription factor of TFEB in regulating autophagy and lysosomal functions and suggest YY1 could be a therapeutic target in cancer treatment.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-HA−Peroxidase antibody, Mouse monoclonal antibody produced in mouse, clone HA-7, purified from hybridoma cell culture