Nuclear Receptor Nur77 Facilitates Melanoma Cell Survival under Metabolic Stress by Protecting Fatty Acid Oxidation.
Molecular cell (2018-02-06)
Xiao-Xue Li, Zhi-Jing Wang, Yu Zheng, Yun-Feng Guan, Peng-Bo Yang, Xiang Chen, Cong Peng, Jian-Ping He, Yuan-Li Ai, Sheng-Fu Wu, Kun-Yi Chien, Qiao Wu, Hang-Zi Chen
RESUMEN
Fatty acid oxidation (FAO) is crucial for cells to overcome metabolic stress by providing ATP and NADPH. However, the mechanism by which FAO is regulated in tumors remains elusive. Here we show that Nur77 is required for the metabolic adaptation of melanoma cells by protecting FAO. Glucose deprivation activates ERK2 to phosphorylate and induce Nur77 translocation to the mitochondria, where Nur77 binds to TPβ, a rate-limiting enzyme in FAO. Although TPβ activity is normally inhibited by oxidation under glucose deprivation, the Nur77-TPβ association results in Nur77 self-sacrifice to protect TPβ from oxidation. FAO is therefore able to maintain NADPH and ATP levels and prevent ROS increase and cell death. The Nur77-TPβ interaction further promotes melanoma metastasis by facilitating circulating melanoma cell survival. This study demonstrates a novel regulatory function of Nur77 with linkage of the FAO-NADPH-ROS pathway during metabolic stress, suggesting Nur77 as a potential therapeutic target in melanoma.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid