Aggregate formation and phosphorylation of neurofilament-L Pro22 Charcot-Marie-Tooth disease mutants.
Human molecular genetics (2006-02-03)
Takahiro Sasaki, Takahiro Gotow, Motoko Shiozaki, Fumika Sakaue, Taro Saito, Jean-Pierre Julien, Yasuo Uchiyama, Shin-Ichi Hisanaga
RESUMEN
Charcot-Marie-Tooth disease (CMT) is the most common inherited peripheral nerve disorder. The causative gene for axonal type CMT2E has been identified as neurofilament light (NF-L) chain. Using cultured cells and in vitro assays, we analyzed the filament formation ability of Pro22 CMT mutant proteins of NF-L, P22S and P22T. NF-L Pro22 mutant proteins formed large aggregates in SW13- cells and cortical neurons and assembled into short twisty threads thinner than 10 nm filaments in vitro. Those threads associated with each other at their ends and entangled into large aggregates, also abnormalities, were detected at steps in oligomer formation. Pro22 mutations abolished Thr21 phosphorylation by cyclin-dependent kinase 5 and external signal regulated kinase, which suppressed filament assembly, but phosphorylation by protein kinase A (PKA) inhibited aggregate formation in vitro and alleviated aggregates in cortical neurons. These results indicate that the Pro22 CMT mutation induces abnormal filament aggregates by disrupting proper oligomer formation and the aggregates are mitigated by phosphorylation with PKA, which makes it a viable target for the development for therapeutics.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-Neurofilament 160 antibody produced in mouse, clone NN18, ascites fluid