• Inicio
  • Resultados de la búsqueda
  • A simultaneous grafting/vinyl polymerization process generates a polycationic surface for enhanced antibacterial activity of bacterial cellulose.
A simultaneous grafting/vinyl polymerization process generates a polycationic surface for enhanced antibacterial activity of bacterial cellulose.
International journal of biological macromolecules (2019-12-10)
Haiyan Liu, Yang Hu, Yongjun Zhu, Xiuping Wu, Xin Zhou, Haobo Pan, Shu Chen, Pengfei Tian
RESUMEN
Bacterial cellulose (BC) is a biosynthesized carbohydrate polymer with excellent biocompatibility and water holding capability. However, it lacks an inherent antibacterial activity that has limited its in-depth biomedical applications. This study investigated a novel strategy of adopting a simultaneous process to chemically anchor a quaternary ammonium salt (R-N(CH3)+) with a special vinyl group (2-methacryloyloxyethyl trimethylammonium chloride, METAC) onto the BC, and meanwhile, enhance the density of (R-N(CH3)+) via free radical vinyl polymerization. The results have confirmed the transition of BC surface from a negatively-charged surface to a polycationic surface via such a simultaneous reaction. As compared to chitin film (a representative of R-NH3+), the resulting METAC-grafted BC (a representative of high-density R- N(CH3)+) acquired excellent water absorbability (40 times of dry weight of the BC), 99% antibacterial activity against Escherichia coli and Staphylococcus aureus, a satisfactory in-vitro biocompatibility, and a better in-vivo wound healing outcome with an excellent in-vivo antibacterial efficacy. This study has exhibited potential in utilizing a facile method to prepare a bio-safe, adaptive antibacterial surface for various biomedical applications.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Chitin from shrimp shells, practical grade, powder
Sigma-Aldrich
[2-(Methacryloyloxy)ethyl]trimethylammonium chloride solution, 75 wt. % in H2O
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥98% (titration)
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder