A phosphorylation-based switch controls TAA1-mediated auxin biosynthesis in plants.
Nature communications (2020-02-06)
Qian Wang, Guochen Qin, Min Cao, Rong Chen, Yuming He, Liyuan Yang, Zhejun Zeng, Yongqiang Yu, Yangtao Gu, Weiman Xing, W Andy Tao, Tongda Xu
RESUMEN
Auxin determines the developmental fate of plant tissues, and local auxin concentration is precisely controlled. The role of auxin transport in modulating local auxin concentration has been widely studied but the regulation of local auxin biosynthesis is less well understood. Here, we show that TRYPTOPHAN AMINOTRANSFERASE OF ARABIDOPSIS (TAA1), a key enzyme in the auxin biosynthesis pathway in Arabidopsis thaliana is phosphorylated at Threonine 101 (T101). T101 phosphorylation status can act as an on/off switch to control TAA1-dependent auxin biosynthesis and is required for proper regulation of root meristem size and root hair development. This phosphosite is evolutionarily conserved suggesting post-translational regulation of auxin biosynthesis may be a general phenomenon. In addition, we show that auxin itself, in part via TRANS-MEMBRANE KINASE 4 (TMK4), can induce T101 phosphorylation of TAA1 suggesting a self-regulatory loop whereby local auxin signalling can suppress biosynthesis. We conclude that phosphorylation-dependent control of TAA1 enzymatic activity may contribute to regulation of auxin concentration in response to endogenous and/or external cues.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Triton X-100, laboratory grade
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, ≥98%
Millipore
Glutathione−Agarose, lyophilized powder
Sigma-Aldrich
Cantharidin
Sigma-Aldrich
Anti-HA−Peroxidase antibody, Mouse monoclonal antibody produced in mouse, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
L-Kynurenine sulfate salt, crystalline