• Inicio
  • Resultados de la búsqueda
  • An F-Box Protein, Mdm30, Interacts with TREX Subunit Sub2 To Regulate Cellular Abundance Cotranscriptionally in Orchestrating mRNA Export Independently of Splicing and Mitochondrial Function.
An F-Box Protein, Mdm30, Interacts with TREX Subunit Sub2 To Regulate Cellular Abundance Cotranscriptionally in Orchestrating mRNA Export Independently of Splicing and Mitochondrial Function.
Molecular and cellular biology (2020-01-15)
Jannatul Ferdoush, Rwik Sen, Geetha Durairaj, Priyanka Barman, Amala Kaja, Shalini Guha, Sukesh R Bhaumik
RESUMEN
Although an F-box protein, Mdm30, is found to regulate ubiquitylation of the Sub2 component of TREX (transcription-export) complex for proteasomal degradation in stimulation of mRNA export, it remains unknown whether such ubiquitin-proteasome system (UPS) regulation of Sub2 occurs cotranscriptionally via its interaction with Mdm30. Further, it is unclear whether impaired UPS regulation of Sub2 in the absence of Mdm30 alters mRNA export via splicing defects of export factors and/or mitochondrial dynamics/function, since Sub2 controls mRNA splicing and Mdm30 regulates mitochondrial aggregation. Here, we show that Mdm30 interacts with Sub2, and temporary shutdown of Mdm30 enhances Sub2's abundance and impairs mRNA export. Likewise, Sub2's abundance is increased following transcriptional inhibition. These results support Mdm30's direct role in regulation of Sub2's cellular abundance in a transcription-dependent manner. Consistently, the chromatin-bound Sub2 level is increased in the absence of Mdm30. Further, we find that Mdm30 does not facilitate splicing of export factors. Moreover, Mdm30 does not have a dramatic effect on mitochondrial respiration/function, and mRNA export occurs in the absence of Fzo1, which is required for mitochondrial dynamics/respiration. Collective results reveal that Mdm30 interacts with Sub2 for proteasomal degradation in a transcription-dependent manner to promote mRNA export independently of splicing or mitochondrial function, thus advancing our understanding of mRNA export.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-HA−Peroxidase antibody, Mouse monoclonal antibody produced in mouse, clone HA-7, purified from hybridoma cell culture