Filling of Nanometric Pores with Polymer by Initiated Chemical Vapor Deposition.
Macromolecular rapid communications (2020-06-11)
Manon Van-Straaten, Amal Ben Hadj Mabrouk, Marc Veillerot, Christophe Licitra, Franck D'Agosto, Vincent Jousseaume
RESUMEN
The integration of porous thin films using microelectronic compatible processes sometimes requires the protection of the interior of the pores during the critical integration steps. In this paper, the polymerization of neo-pentyl methacrylate (npMA) is performed via initiated chemical vapor deposition (iCVD) on a porous organosilicate (SiOCH) and on a dense SiOCH. The characterizations by Fourier-transform infrared spectroscopy, spectroscopic ellipsometry, and time-of-flight secondary ion mass spectrometry of the different stacks show that iCVD is a powerful technique to polymerize npMA in the nanometric pores and thus totally fill them with a polymer. The study of the pore filling for very short iCVD durations shows that the polymerization in the pores is complete in less than ten seconds and is uniform in depth. Then, the poly(npMA) film growth continues on top of the filled SiOCH layer. These characteristics make iCVD a straightforward and very promising alternative to other infiltration techniques in order to fill the porosity of microporous thin films.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Luperox® DI, tert-Butyl peroxide, 98%
Sigma-Aldrich
2-Chloroethyl acrylate, contains >100 ppm MEHQ as inhibitor, 97%