The Lysine Specific Demethylase-1 Negatively Regulates the COL9A1 Gene in Human Articular Chondrocytes.
International journal of molecular sciences (2020-09-04)
Anne-Laure Durand, Alexandre Dufour, Elisabeth Aubert-Foucher, Christine Oger-Desfeux, Marielle Pasdeloup, Sebastien Lustig, Elvire Servien, Gualter Vaz, Emeline Perrier-Groult, Frederic Mallein-Gerin, Jerome E Lafont
RESUMEN
Osteoarthritis (OA) is a degenerative disease of the joints which is associated with an impaired production of the cartilage matrix by the chondrocytes. Here, we investigated the role of Lysine-Specific Demethylase-1 (LSD1), a chromatin remodeling enzyme whose role in articular chondrocytes was previously associated with a catabolic activity and which is potentially involved during OA. Following a loss of function strategy and RNA sequencing analysis, we detail the genes which are targeted by LSD1 in human articular chondrocytes and identify COL9A1, a gene encoding the α1 chain of the cartilage-specific type IX collagen, as negatively regulated by LSD1. We show that LSD1 interacts with the transcription factor SOX9 and is recruited to the promoter of COL9A1. Interestingly, we observe that OA cartilage displays stronger LSD1 immunostaining compared with normal, and we demonstrate that the depletion of LSD1 in OA chondrocytes prevents the decrease in COL9A1 following Il-1β treatment. These results suggest LSD1 is a new regulator of the anabolic activity of articular chondrocytes potentially destabilizing the cartilage matrix, since it negatively regulates COL9A1, a gene encoding a crucial anchoring collagen molecule. This newly identified role played by LSD1 may thus participate in the alteration of the cartilage matrix during OA.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-β-Tubulin I+II antibody produced in mouse, clone JDR.3B8, ascites fluid
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.