Nuclear entry of hyperbranched polylysine nanoparticles into cochlear cells.
International journal of nanomedicine (2011-04-07)
Weikai Zhang, Ya Zhang, Marian Löbler, Klaus-Peter Schmitz, Aqeel Ahmad, Ilmari Pyykkö, Jing Zou
RESUMEN
Gene therapy is a potentially effective therapeutic modality for treating sensorineural hearing loss. Nonviral gene delivery vectors are expected to become extremely safe and convenient, and nanoparticles are the most promising types of vectors. However, infrequent nuclear localization in the cochlear cells limits their application for gene therapy. This study aimed to investigate the potential nuclear entry of hyperbranched polylysine nanoparticles (HPNPs) for gene delivery to cochlear targets. Rat primary cochlear cells and cochlear explants generated from newborn rats were treated with different concentrations of HPNPs. For the in vivo study, HPNPs were administered to the rats' round window membranes. Subcellular distribution of HPNPs in different cell populations was observed with confocal microscope 24 hours after administration. Nuclear entry was observed in various cochlear cell types in vitro and in vivo. In the primary cochlear cell culture, concentration-dependent internalization was observed. In the cochlear organotypic culture, abundant HPNPs were found in the modiolus, including the spiral ganglion, organ of Corti, and lateral wall tissues. In the in vivo study, a gradient distribution of HPNPs through different layers of the round window membrane was observed. HPNPs were also distributed in the cells of the middle ear tissue. Additionally, efficient internalization of HPNPs was observed in the organ of Corti and spiral ganglion cells. In primary cochlear cells, HPNPs induced higher transfection efficiency than did Lipofectamine(™). These results suggest that HPNPs are potentially an ideal carrier for gene delivery into the cochlea.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Phosphate Buffered Saline with 0.05% TWEEN® 20, pH 7.4
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Phosphate buffered saline, BioPerformance Certified, pH 7.4
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Phosphate Buffered Saline, 10× PBS for Western blots and IP
SAFC
L-Glutamine
Sigma-Aldrich
Phosphate buffered saline, pH 7.2 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 5 L solutions
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Phosphate buffered saline, BioUltra, pH 7.4 ( in solution), contains TWEEN 20, tablet
Sigma-Aldrich
Phosphate buffered saline, pH 7.4, contains BSA, powder
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Phosphate buffered saline, BioUltra, washing buffer for peroxidase conjugates in Western Blotting, 10x concentrate
Sigma-Aldrich
Phosphate buffered saline, BioUltra, solution
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Phosphate Buffered Saline with 3% Non-Fat Milk, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, Autoclaved, pH 7.2 (25 °C)
Sigma-Aldrich
Epidermal Growth Factor from mouse, EGF
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphate buffered saline, pH 7.6 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, with 5% nonfat milk, powder (dry milled), pH 7.3
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Glutamine