Intellectual disability-associated gene ftsj1 is responsible for 2'-O-methylation of specific tRNAs.
EMBO reports (2020-06-20)
Jing Li, Yan-Nan Wang, Bei-Si Xu, Ya-Ping Liu, Mi Zhou, Tao Long, Hao Li, Han Dong, Yan Nie, Peng R Chen, En-Duo Wang, Ru-Juan Liu
RESUMEN
tRNA modifications at the anti-codon loop are critical for accurate decoding. FTSJ1 was hypothesized to be a human tRNA 2'-O-methyltransferase. tRNAPhe (GAA) from intellectual disability patients with mutations in ftsj1 lacks 2'-O-methylation at C32 and G34 (Cm32 and Gm34). However, the catalytic activity, RNA substrates, and pathogenic mechanism of FTSJ1 remain unknown, owing, in part, to the difficulty in reconstituting enzymatic activity in vitro. Here, we identify an interacting protein of FTSJ1, WDR6. For the first time, we reconstitute the 2'-O-methylation activity of the FTSJ1-WDR6 complex in vitro, which occurs at position 34 of specific tRNAs with m1 G37 as a prerequisite. We find that modifications at positions 32, 34, and 37 are interdependent and occur in a hierarchical order in vivo. We also show that the translation efficiency of the UUU codon, but not the UUC codon decoded by tRNAPhe (GAA), is reduced in ftsj1 knockout cells. Bioinformatics analysis reveals that almost 40% of the high TTT-biased genes are related to brain/nervous functions. Our data potentially enhance our understanding of the relationship between FTSJ1 and nervous system development.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal antibody produced in mouse, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution