Possible involvement of radical intermediates in the inhibition of cysteine proteases by allenyl esters and amides.
Bioorganic & medicinal chemistry letters (2008-10-28)
Yoshio Takeuchi, Tomoya Fujiwara, Yoshihito Shimone, Hideki Miyataka, Toshio Satoh, Kenneth L Kirk, Hitoshi Hori
RESUMEN
In order to investigate crystallographically the mechanism of inhibition of cysteine protease by alpha-methyl-gamma,gamma-diphenylallenecarboxylic acid ethyl ester 3, a cysteine protease inhibitor having in vivo stability, we synthesized N-(alpha-methyl-gamma,gamma-diphenylallenecarbonyl)-L-phenylalanine ethyl ester 4. Reaction of 4 with thiophenol, the SH group of which has similar pK(a) value to that of cysteine protease, produced oxygen-mediated radical adducts 6 and 7 in ambient air but did not proceed under oxygen-free conditions. Catalytic activities of two thiol enzymes including cathepsin B were also lowered in the absence of oxygen. These results suggest that cysteine protease can act through an oxygen-dependent radical mechanism.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
SAFC
L-Cysteine
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Thiophenol, 97%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
Thiophenol, ≥99%
Sigma-Aldrich
Famotidine