Merck
Antioxidant and antibacterial studies of arylazopyrazoles and arylhydrazonopyrazolones containing coumarin moiety.
European journal of medicinal chemistry (2009-08-04)
P Manojkumar, T K Ravi, S Gopalakrishnan
RESUMEN
A series of novel 1-(4-methylcoumarinyl-7-oxyacetyl)-3,5-dimethyl-4(arylazo)pyrazoles and 1-(4-methylcoumarinyl-7-oxyacetyl)-3-methyl-4-(substituted phenyl) hydrazono-2-pyrazolin-5-ones were synthesised and evaluated for antibacterial and antioxidant activities. Compounds 3b, 3g, 5b, 5d and 5g showed good antibacterial activity and compound 5e was found to be the most active antioxidant in the series, and thus represent a new class of promising lead compounds.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, 99%