  • Search Results
  • Effect of Rooibos (Aspalathus linearis) extract on atorvastatin-induced toxicity in C3A liver cells.
Effect of Rooibos (Aspalathus linearis) extract on atorvastatin-induced toxicity in C3A liver cells.
Journal of cellular physiology (2020-05-28)
Danielle A Millar, Sandra Bowles, Shantal Lynn Windvogel, Johan Louw, Christo J F Muller
ABSTRACT
Rooibos (Aspalathus linearis) has various health benefits. Two case studies have associated chronic Rooibos consumption with conventional prescription medications, including atorvastatin (ATV), with hepatotoxicity. Statins act by inhibiting hydroxymethylglutaryl-coenzyme A reductase, a rate-limiting enzyme in cholesterol synthesis. Although rare, statins are potentially hepatotoxic. The aim was to investigate interactions between aspalathin-rich Rooibos extract GRT™ and ATV-induced hepatotoxicity in C3A liver cells cultured with and without palmitate. Effects of co-treatment of GRT + ATV on cell viability, oxidative stress, apoptosis, mitochondrial integrity, and cellular reactive oxygen species (ROS) production were assessed. Significantly increased ROS production was observed in cells exposed to ATV and palmitate. Combination therapy of GRT + ATV also showed significant increases in ROS production. Under palmitate-treated conditions, ATV-induced significant apoptosis which was not ameliorated by GRT + ATV co-treatment. Despite studies purporting hepatoprotection from Rooibos, our study showed that GRT was unable to modulate ATV-induced hepatotoxic effects in this model.
Sigma-Aldrich
2′,7′-Dichlorofluorescin diacetate, ≥97%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Roche
Bovine Serum Albumin Fraction V, from bovine serum