Merck
Electrophysiological evaluation of human induced pluripotent stem cell-derived cardiomyocytes obtained by different methods.
Stem cell research (2021-01-24)
Chandra Prajapati, Marisa Ojala, Henna Lappi, Katriina Aalto-Setälä, Mari Pekkanen-Mattila
RESUMEN
The human induced pluripotent stem cells (hiPSCs) derived cardiomyocytes (CMs) (hiPSC-CMs) retain the same genetic information as the donor, and they have been shown to faithfully recapitulate the disease phenotypes of various genetic cardiac diseases. The hiPSC-CMs can be utilized in multiple types of studies and in most cases, the functionality of hiPSC-CMs is of interest. For the functional analyses, the hiPSC-CMs need to be manipulated after differentiation, e.g. enriched or dissociated into single-cell stage. For the functional assessments to be reliable and reproducible, the cell culture environment should support the cells in an optimal manner. The aim of the present study was to evaluate the effect of various differentiation methods, as well as coating materials used for the dissociated cells on the functionality of the differentiated hiPSC-CMs. The different protocols not only had different differentiation efficiencies, but they also yielded functionally different hiPSC-CMs. Additionally, the coating material had a major effect on the functionality of the hiPSC-CMs. The results of the present study emphasize that the cardiac differentiation method and the coating material have a major effect on hiPS-CMs' characteristics. Thus, when different hiPSC lines and results obtained in different labs are compared, extra care should be taken to check the conditions when results are compared.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate, ≥95%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
E-4031, ≥98% (HPLC), lyophilized powder
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Anti-Connexin-43 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
4-Aminopyridine, 98%
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
(−)-Blebbistatin, solid, synthetic
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Nimodipine
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
IWR-1, ≥98% (HPLC)