Human DICER helicase domain recruits PKR and modulates its antiviral activity.
PLoS pathogens (2021-05-14)
Thomas C Montavon, Morgane Baldaccini, Mathieu Lefèvre, Erika Girardi, Béatrice Chane-Woon-Ming, Mélanie Messmer, Philippe Hammann, Johana Chicher, Sébastien Pfeffer
RESUMEN
The antiviral innate immune response mainly involves type I interferon (IFN) in mammalian cells. The contribution of the RNA silencing machinery remains to be established, but several recent studies indicate that the ribonuclease DICER can generate viral siRNAs in specific conditions. It has also been proposed that type I IFN and RNA silencing could be mutually exclusive antiviral responses. In order to decipher the implication of DICER during infection of human cells with alphaviruses such as the Sindbis virus and Semliki forest virus, we determined its interactome by proteomics analysis. We show that DICER specifically interacts with several double-stranded RNA binding proteins and RNA helicases during viral infection. In particular, proteins such as DHX9, ADAR-1 and the protein kinase RNA-activated (PKR) are enriched with DICER in virus-infected cells. We demonstrate that the helicase domain of DICER is essential for this interaction and that its deletion confers antiviral properties to this protein in an RNAi-independent, PKR-dependent, manner.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Monoclonal Anti-γ-Tubulin antibody produced in mouse, clone GTU-88, ascites fluid