Domain-swap polymerization drives the self-assembly of the bacterial flagellar motor.
Nature structural & molecular biology (2016-02-09)
Matthew A B Baker, Robert M G Hynson, Lorraine A Ganuelas, Nasim Shah Mohammadi, Chu Wai Liew, Anthony A Rey, Anthony P Duff, Andrew E Whitten, Cy M Jeffries, Nicolas J Delalez, Yusuke V Morimoto, Daniela Stock, Judith P Armitage, Andrew J Turberfield, Keiichi Namba, Richard M Berry, Lawrence K Lee
RESUMEN
Large protein complexes assemble spontaneously, yet their subunits do not prematurely form unwanted aggregates. This paradox is epitomized in the bacterial flagellar motor, a sophisticated rotary motor and sensory switch consisting of hundreds of subunits. Here we demonstrate that Escherichia coli FliG, one of the earliest-assembling flagellar motor proteins, forms ordered ring structures via domain-swap polymerization, which in other proteins has been associated with uncontrolled and deleterious protein aggregation. Solution structural data, in combination with in vivo biochemical cross-linking experiments and evolutionary covariance analysis, revealed that FliG exists predominantly as a monomer in solution but only as domain-swapped polymers in assembled flagellar motors. We propose a general structural and thermodynamic model for self-assembly, in which a structural template controls assembly and shapes polymer formation into rings.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-HA−Peroxidase antibody, Mouse monoclonal antibody produced in mouse, clone HA-7, purified from hybridoma cell culture