A monoclonal antibody against the type II isotype of beta-tubulin. Preparation of isotypically altered tubulin.
The Journal of biological chemistry (1988-02-25)
A Banerjee, M C Roach, K A Wall, M A Lopata, D W Cleveland, R F Ludueña
RESUMEN
Mammalian brain tubulin consists of several isotypes of alpha and beta subunits that separate on polyacrylamide gels into three electrophoretic classes, designated alpha, beta 1, and beta 2. It has not been possible hitherto to resolve the different isotypes in a functional form. To this end, we have now isolated a monoclonal antibody, using as an immunogen a chemically synthesized peptide corresponding to the carboxyl-terminal sequence of the major tubulin isotype (type II) found in the beta 1-tubulin electrophoretic fraction. The antibody binds to beta 1 but not to alpha or beta 2. When pure tubulin from bovine brain is passed through an immunoaffinity column made from the anti-type II antibody, the tubulin that elutes in the unbound fraction is enriched greatly for the beta 2 electrophoretic variant. The tubulin that binds to the column appears to contain only alpha and beta 1, not beta 2. When these tubulin fractions are characterized by immunoblotting using the anti-type II antibody, the antibody binds only to the beta 1 band in the bound fraction, not to the beta 1 band in the unbound fraction. Using polyclonal antibodies generated against the carboxyl-termini of types I, III, and IV, we demonstrate that the beta 1 electrophoretic species is comprised of isotypes I, II, and IV, whereas the beta 2 variant is comprised exclusively of type III beta-tubulin. Further, we calculate that beta-tubulin in purified bovine brain tubulin is comprised of 3% type I, 58% type II, 25% type III, and 13% type IV tubulins.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Tubulin I+II antibody produced in mouse, clone JDR.3B8, ascites fluid
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.