Leukocyte cell-derived chemotaxin 2 is a zinc-binding protein.
FEBS letters (2013-01-29)
Akinori Okumura, Takehiro Suzuki, Hideyuki Miyatake, Tomoya Okabe, Yuki Hashimoto, Takuya Miyakawa, Hai Zheng, Hiroyuki Unoki-Kubota, Hideaki Ohno, Naoshi Dohmae, Yasushi Kaburagi, Yoshitsugu Miyazaki, Masaru Tanokura, Satoshi Yamagoe
RESUMEN
Leukocyte cell-derived chemotaxin 2 (LECT2) is a secreted hepatic protein that has been associated with several physiological activities. LECT2 belongs to the peptidase M23 family, suggesting that it is a zinc-binding protein. To test this possibility, electrospray ionization mass spectrometry and X-ray absorption fine-structure analysis were performed. Results of these experiments indicated that recombinant mouse LECT2 produced by an animal cell line contains a zinc atom. Furthermore, the recombinant LECT2 was found to be self-oligomerized by disulfide bonds in vitro, but this was suppressed by addition of zinc. These results indicated that zinc stabilizes the LECT2 structure.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Lysostaphin from Staphylococcus simulans, recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, BioUltra, ≥97% (SDS-PAGE), Protein 40-60 % by biuret, ≥2,000 units/mg protein
SAFC
L-Cystine
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, free of DNA contaminants, suitable for Microbiome research, lyophilized powder, ≥500 units/mg protein
SAFC
L-Cystine
Supelco
L-Cystine, certified reference material, TraceCERT®
Cystine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, aseptically filled
Sigma-Aldrich
L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%