Merck
Role of inhibition in respiratory pattern generation.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-03-29)
Wiktor A Janczewski, Alexis Tashima, Paul Hsu, Yan Cui, Jack L Feldman
RESUMEN
Postsynaptic inhibition is a key element of neural circuits underlying behavior, with 20-50% of all mammalian (nongranule) neurons considered inhibitory. For rhythmic movements in mammals, e.g., walking, swimming, suckling, chewing, and breathing, inhibition is often hypothesized to play an essential rhythmogenic role. Here we study the role of fast synaptic inhibitory neurotransmission in the generation of breathing pattern by blocking GABA(A) and glycine receptors in the preBötzinger complex (preBötC), a site essential for generation of normal breathing pattern, and in the neighboring Bötzinger complex (BötC). The breathing rhythm continued following this blockade, but the lung inflation-induced Breuer-Hering inspiratory inhibitory reflex was suppressed. The antagonists were efficacious, as this blockade abolished the profound effects of the exogenously applied GABA(A) receptor agonist muscimol or glycine, either of which under control conditions stopped breathing in vagus-intact or vagotomized, anesthetized, spontaneously breathing adult rats. In vagotomized rats, GABA(A)ergic and glycinergic antagonists had little, if any, effect on rhythm. The effect in vagus-intact rats was to slow the rhythm to a pace equivalent to that seen after suppression of the aforementioned Breuer-Hering inflation reflex. We conclude that postsynaptic inhibition within the preBötC and BötC is not essential for generation of normal respiratory rhythm in intact mammals. We suggest the primary role of inhibition is in shaping the pattern of respiratory motor output, assuring its stability, and in mediating reflex or volitional apnea, but not in the generation of rhythm per se.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Muscimol, powder
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Strychnine
Supelco
Strychnine, PESTANAL®, analytical standard
Sigma-Aldrich
(+)-Bicuculline, ≥97.0% (TLC)
Sigma-Aldrich
DAPI, for nucleic acid staining