  • Search Results
  • Secretory phospholipase A2-IID is an effector molecule of CD4+CD25+ regulatory T cells.
Secretory phospholipase A2-IID is an effector molecule of CD4+CD25+ regulatory T cells.
Proceedings of the National Academy of Sciences of the United States of America (2009-07-01)
Caroline E von Allmen, Nicole Schmitz, Monika Bauer, Heather J Hinton, Michael O Kurrer, Regula B Buser, Myriam Gwerder, Simone Muntwiler, Tim Sparwasser, Roger R Beerli, Martin F Bachmann
ABSTRACT
Suppression by natural CD4(+)CD25(+) regulatory T cells (Tregs) is one mechanism by which tolerance is maintained. However, the way in which Tregs mediate suppression is not well understood. Here, we show that secreted phospholipase A2 (sPLA2)-IID is selectively produced by Tregs. sPLA2-IID is a potent mediator of Treg function, because it strongly suppressed proliferation of CD4(+) and CD8(+) T cells in vitro and in vivo in a manner independent of its catalytic activity. Furthermore, sPLA2-IID promoted the differentiation of Tregs, presumably via attenuating signaling through the PI3K/Akt/mammalian target of rapamycin pathway. Importantly, administration of a sPLA2-IID-Fc fusion protein inhibited disease development in murine models of colitis and multiple sclerosis, suggesting that sPLA2-IID's immunosuppressive function might be exploited therapeutically.
Sigma-Aldrich
Paraformaldehyde, meets analytical specification of DAC, 95.0-100.5%
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Paraformaldehyde, prilled, 95%
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Supelco
Rapamycin, VETRANAL®, analytical standard