• Inicio
  • Resultados de la búsqueda
  • Synthesis, spectral and thermal studies of the newly hydrogen bonded charge transfer complex of o-phenylenediamine with pi acceptor picric acid.
Synthesis, spectral and thermal studies of the newly hydrogen bonded charge transfer complex of o-phenylenediamine with pi acceptor picric acid.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2010-07-03)
Ishaat M Khan, Afaq Ahmad
RESUMEN
Newly proton or charge transfer complex [(OPDH)(+)(PA)(-)] was synthesized by the reaction of the donor, o-phenylenediamine (OPD) with acceptor, 2,4,6-trinitrophenol (PAH). The chemical reaction has occurred via strong hydrogen bonding followed by migration of proton from acceptor to donor. UV-vis, (1)H NMR and FTIR spectra, in addition to the thermal and elemental analysis were used to confirm the proposed occurrence of the chemical reaction and to investigate the newly synthesized solid CT complex. The stoichiometry of the CT complex was found to be 1:1. The formation constant and molar extinction coefficient of the CT complex were evaluated by the Benesi-Hildebrand equation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, peroxidase substrate
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%