Copper-catalyzed synthesis of quinoxalines with o-phenylenediamine and terminal alkyne in the presence of bases.
Organic letters (2011-08-03)
Wen Wang, Yongwen Shen, Xu Meng, Mingming Zhao, Yongxin Chen, Baohua Chen
RESUMEN
A novel way of synthesizing quinoxalines efficiently through cyclization of o-phenylenediamine and terminal alkyne by Cu(II) and bases is developed. This reaction proceeds smoothly to give the products in moderate to good yields.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, peroxidase substrate
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%