• Inicio
  • Resultados de la búsqueda
  • Petalonia binghamiae extract and its constituent fucoxanthin ameliorate high-fat diet-induced obesity by activating AMP-activated protein kinase.
Petalonia binghamiae extract and its constituent fucoxanthin ameliorate high-fat diet-induced obesity by activating AMP-activated protein kinase.
Journal of agricultural and food chemistry (2012-03-10)
Seong-Il Kang, Hye-Sun Shin, Hyo-Min Kim, Seon-A Yoon, Seung-Woo Kang, Jeong-Hwan Kim, Hee-Chul Ko, Se-Jae Kim
RESUMEN
In this study, we investigated the antiobesity properties of Petalonia binghamiae extract (PBE) in mice in which obesity was induced with a high-fat diet (HFD). PBE administration (150 mg/kg/day) for 70 days decreased body weight gain, adipose tissue weight, and the serum triglyceride level in mice fed a HFD. PBE reduced serum levels of glutamic pyruvic transaminase and glutamic oxaloacetic transaminase as well as the accumulation of lipid droplets in the liver. PBE restored the HFD-induced decrease in phosphorylation of AMP-activated protein kinase (AMPK) and acetyl-CoA carboxylase (ACC) in epididymal adipose tissue. PBE increased the phosphorylation of AMPK and ACC and decreased the expression of SREBP1c in mature 3T3-L1 adipocytes. In addition, we further explored the active compound responsible for AMPK activation by PBE in 3T3-L1 adipocytes. Fucoxanthin isolated from PBE increased the phosphorylation of AMPK and ACC with increasing LKB1 phosphorylation in mature 3T3-L1 adipocytes. Taken together, these data suggest that PBE (or fucoxanthin) exert improving effects on HFD-induced obesity by promoting β-oxidation and reducing lipogenesis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Fucoxanthin, carotenoid antioxidant