• Inicio
  • Resultados de la búsqueda
  • Severe hyperphosphatemia after administration of sodium-phosphate containing laxatives in children: case series and systematic review of literature.
Severe hyperphosphatemia after administration of sodium-phosphate containing laxatives in children: case series and systematic review of literature.
Pediatric surgery international (2012-07-24)
Hannah N Ladenhauf, Ottokar Stundner, Florian Spreitzhofer, Stefan Deluggi
RESUMEN
Sodium phosphate-containing laxatives are commonly used as first-line treatment option for constipation in children and adolescents. Hyperphosphatemia is an infrequent, but potentially life-threatening complication of laxative application. We report a case series of three children exhibiting severe hyperphosphatemia and hypocalcemia after utilization of sodium phosphate-containing laxatives, necessitating intensive care services in two of three cases. Additionally, we reviewed 32 case reports of similar occurrences. We identified 28 publications on the subject dating from 1968 to 2010. Mean age of all children in reports was 2.83 years; sex was approximately equally distributed. While 18 patients suffered from either pre-existing gastrointestinal comorbidity or other major systemic disease, no or only unrelated, minor conditions were present in ten children. One-third of patients received laxatives repeatedly before the incident. Findings associated with hyperphosphatemia include lethargy, dizziness, stiffness, tachypnea, tachycardia and severe dehydration in almost all cases, and tetany, carpopedal spasm, and prolonged QT interval in a subset. While about 80% of children recovered without residual findings, three deceased and one incurred persistent hypoxic brain damage. Physicians should be alerted to the possibility of phosphate toxicity in children and adolescents treated with laxatives.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ACS reagent, 98.0-102.0%
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)