Consensus guidelines for the safe prescription and administration of oral bowel-cleansing agents.
Gut (2012-07-31)
Andrew Connor, Damian Tolan, Stephen Hughes, Nick Carr, Charles Tomson
RESUMEN
Oral bowel-cleansing preparations are used before colonic surgery and endoscopic and radiological assessment of the intestine to minimise faecal contamination. In February 2009, the UK National Patient Safety Agency issued a Rapid Response Report highlighting the potential risk of harm associated with the use of these preparations and instructing local NHS Trusts to implement safeguards to reduce this risk. This guidance has been prepared to help NHS Trusts to respond to these concerns, as the risk of complications is influenced by both individual patient risk factors and the choice of bowel preparation, for which definitive guidance was not previously available. This document provides an outline of the different available oral bowel-cleansing agents and the complications that may arise. This is followed by recommendations for their use in different patient groups and circumstances. The recommendations are based on consensus between the authors, each of whom circulated drafts to members of their specialist society. The evidence for these recommendations has been assessed using the modified GRADE system. The recommendations cover the choice, administration and complications (relative and absolute) of the different oral bowel-cleansing agents, with specific guidance provided for different patient groups.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ACS reagent, 98.0-102.0%
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)