Persulfate oxidation regeneration of granular activated carbon: reversible impacts on sorption behavior.
Chemosphere (2012-08-28)
Andy Hutson, Saebom Ko, Scott G Huling
RESUMEN
Chemical oxidation regeneration of granular activated carbon (GAC) is a developing technology that can be carried out utilizing thermally-activated persulfate. During chemical regeneration of GAC, aggressive oxidative conditions lead to high acidity (pH<2) and the accumulation of sodium persulfate residuals in the GAC. In this study, we investigated the impact of chemical oxidation on the sorption characteristics of methyl-tert butyl ether (MTBE) in GAC. Loss of MTBE sorption was measured in thermally-activated persulfate regenerated GAC. The accumulation of sulfur was partially responsible for the blockage of sorption sites, but sorption loss was amplified under oxidizing and acidic conditions and attributed to the formation of acidic surface oxides and enhanced electrostatic attraction and accumulation of SO(4)(2-) in GAC. Raising the pH in the GAC slurry resulted in the removal of the residual sulfate and improved MTBE sorption indicating that the mechanisms responsible for MTBE sorption loss were reversible. These results establish baseline conditions and parameters that can be used to optimize pilot- and full-scale deployment of thermally-activated persulfate regeneration of GAC.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Supelco
tert-Butyl methyl ether, suitable for HPLC, ≥99.8% (GC)