Molecular mechanism of the assembly of an acid-sensing receptor ion channel complex.
Nature communications (2012-12-06)
Yong Yu, Maximilian H Ulbrich, Ming-Hui Li, Scott Dobbins, Wei K Zhang, Liang Tong, Ehud Y Isacoff, Jian Yang
RESUMEN
Polycystic kidney disease (PKD) family proteins associate with transient receptor potential (TRP) channel family proteins to form functionally important complexes. PKD proteins differ from known ion channel-forming proteins and are generally thought to act as membrane receptors. Here we find that PKD1L3, a PKD protein, functions as a channel-forming subunit in an acid-sensing heteromeric complex formed by PKD1L3 and TRPP3, a TRP channel protein. Single amino-acid mutations in the putative pore region of both proteins alter the channel's ion selectivity. The PKD1L3/TRPP3 complex in the plasma membrane of live cells contains one PKD1L3 and three TRPP3. A TRPP3 C-terminal coiled-coil domain forms a trimer in solution and in crystal, and has a crucial role in the assembly and surface expression of the PKD1L3/TRPP3 complex. These results demonstrate that PKD subunits constitute a new class of channel-forming proteins, enriching our understanding of the function of PKD proteins and PKD/TRPP complexes.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dimethylamine hydrochloride, 99%
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Dimethylamine solution, 2.0 M in THF
Sigma-Aldrich
Dimethylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Trimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Trimethylamine hydrochloride, 98%
Sigma-Aldrich
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
Dimethylamine solution, 2.0 M in methanol
Sigma-Aldrich
Dimethylamine solution, purum, 33% in absolute ethanol (~5.6 M)
Sigma-Aldrich
Dimethylamine hydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
Dimethylamine, purum, ≥99.0%
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in propylene glycol