Comparison of the performance of three ion mobility spectrometers for measurement of biogenic amines.
Analytica chimica acta (2012-12-19)
Zeev Karpas, Ana V Guamán, Antonio Pardo, Santiago Marco
RESUMEN
The performance of three different types of ion mobility spectrometer (IMS) devices: GDA2 with a radioactive ion source (Airsense, Germany), UV-IMS with a photo-ionization source (G.A.S. Germany) and VG-Test with a corona discharge source (3QBD, Israel) was studied. The gas-phase ion chemistry in the IMS devices affected the species formed and their measured reduced mobility values. The sensitivity and limit of detection for trimethylamine (TMA), putrescine and cadaverine were compared by continuous monitoring of a stream of air with a given concentration of the analyte and by measurement of headspace vapors of TMA in a sealed vial. Preprocessing of the mobility spectra and the effectiveness of multivariate curve resolution techniques (MCR-LASSO) improved the accuracy of the measurements by correcting baseline effects and adjusting for variations in drift time as well as enhancing the signal to noise ratio and deconvolution of the complex data matrix to their pure components. The limit of detection for measurement of the biogenic amines by the three IMS devices was between 0.1 and 1.2ppm (for TMA with the VG-Test and GDA, respectively) and between 0.2 and 0.7ppm for putrescine and cadaverine with all three devices. Considering the uncertainty in the LOD determination there is almost no statistically significant difference between the three devices although they differ in their operating temperature, ionization method, drift tube design and dopant chemistry. This finding may have general implications on the achievable performance of classic IMS devices.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1,4-Diaminobutane, 99%
Sigma-Aldrich
Trimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Trimethylamine hydrochloride, 98%
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in propylene glycol