Interactions of disulfide-constrained cyclic tetrapeptides with Cu(2+).
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2013-01-24)
Liyun Zhang, Zhaofeng Luo, Lidong Zhang, Liangyuan Jia, Lifang Wu
RESUMEN
The purpose of this work is to characterize the interactions of two disulfide-constrained cyclic tetrapeptides [c(Ac-Cys-Pro-Phe-Cys-NH(2)), SS1; c(Ac-Cys-Pro-Gly-Cys-NH(2)), SS2] with Cu(2+) ions in order to facilitate the design of cyclic peptides as sensors for metal ions. The Cu(2+)-peptide complex cations at m/z 569.1315 for Cu(2+)-SS1 and m/z 479.0815 for Cu(2+)-SS2 were detected by mass spectrometry. The gas-phase fragmentation of the Cu(2+)-peptide complexes was studied by collision-induced dissociation and suggests the atoms involved in the coordination. Cu(2+) ion binds to a single SS1 or SS2 with K (d(app)) of 0.57 ± 0.02 and 0.55 ± 0.01 μM, respectively. Isothermal titration calorimetry data indicate both enthalpic and entropic contributions for the binding of Cu(2+) ion to SS1 and SS2. The characteristic wavenumber of 947 cm(-1) and the changes at 1,664 and 1,530 cm(-1) in the infrared spectrum suggest that the sulfydryl of cysteine, the carbonyl group, and amide II are involved in the coordination of Cu(2+). The X-ray absorption near-edge structure signal from the Cu(2+)-peptide complex corresponds to the four-coordination structure. The extended X-ray absorption fine structure and electron paramagnetic resonance results demonstrate the Cu(2+) ion is in an S/N/2O coordination environment, and is a distinct type II copper center. Theoretical calculations further demonstrate that Cu(2+) ion binds to SS1 or SS2 in a slightly distorted tetragonal geometry with an S/N/2O environment and the minimum potential energy.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
SAFC
L-Cystine
SAFC
L-Cystine
Supelco
L-Cystine, certified reference material, TraceCERT®
Cystine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%