• Inicio
  • Resultados de la búsqueda
  • Decontamination treatments can increase the prevalence of resistance to antibiotics of Escherichia coli naturally present on poultry.
Decontamination treatments can increase the prevalence of resistance to antibiotics of Escherichia coli naturally present on poultry.
Food microbiology (2013-03-19)
Rosa Capita, Elena Alvarez-Fernández, Esther Fernández-Buelta, Jennifer Manteca, Carlos Alonso-Calleja
RESUMEN
The main objective of this study was to determine the ability of various decontaminants to increase the prevalence of resistance to antibiotics in Escherichia coli populations on poultry. Chicken legs were dipped for 15 min into aqueous solutions (wt/vol) of trisodium phosphate (TSP; 12%), acidified sodium chlorite (ASC; 1200 ppm), ascorbic acid (AA; 2%) or citric acid (CA; 2%), or tap water (control). Samples were analyzed immediately after treatment (day 0) and after five days of storage at 7 ± 1 °C. A total of 250 E. coli isolates (50 from each group of samples; 25 on day 0 and 25 on day 5) were tested against twelve antibiotics of clinical significance by means of a standard disc-diffusion technique. A high prevalence of resistance to antibiotics was observed for E. coli strains from control samples, with three (6.0%) isolates sensitive, three (6.0%) resistant to one antibiotic and 44 (88.0%) isolates resistant to two or more antibiotics. Isolates from control samples had a lower prevalence of resistance than those from treated samples to ampicillin-sulbactam (P < 0.01, samples treated with TSP), amoxicillin-clavulanic acid (P < 0.001, ASC, AA and CA), cephotaxime (P < 0.05, TSP), trimethoprim-sulphamethoxazole (P < 0.05, AA; P < 0.01, CA), tetracycline (P < 0.01, CA), ciprofloxacin (P < 0.001, ASC; P < 0.05, AA; P < 0.01, CA) and nitrofurantoin (P < 0.01, TSP). These results suggest that the chemical decontaminants tested could favor the emergence, selection and/or proliferation of antibiotic-resistant strains in microbial populations on poultry meat.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ACS reagent, 98.0-102.0%
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, meets USP testing specifications
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ≥99.99% trace metals basis