Merck
Transient pearling and vesiculation of membrane tubes under osmotic gradients.
Faraday discussions (2013-06-29)
Jeremy Sanborn, Kamila Oglecka, Rachel S Kraut, Atul N Parikh
RESUMEN
We report the experimental observation of osmotically induced transient pearling instabilities in vesicular membranes. Giant phospholipid vesicles subjected to negative osmotic gradient, which drives the influx of water in to the vesicular interior, produces transient cylindrical protrusions. These protrusions exhibit a remarkable pearling intermediate, which facilitates their subsequent retraction. The pearling front propagates from the distal free end of the protrusion toward the vesicular source and accompanies gradual shortening of the protrusion via pearl-pearl coalescence. Real-time introduction of a positive osmotic gradient, on the other hand, drives vigorous shape fluctuations, which in turn produce cylindrical, prolate- and pear-shaped intermediates presumably due to an increased vesicular area relative to the encapsulated volume. These intermediates transiently produce a pearled state prior to their fission. In both cases, the transient pearling state gives rise to an array of stable spherical daughter vesicles, which may be connected to one another by fine tethers not resolved in our experiments. These results may have implications for self-reproduction in primitive, protein-free, cells.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1,2-Dipalmitoyl-rac-glycero-3-phosphoethanolamine, synthetic, ≥98%
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine, ≥97%
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sucrose, European Pharmacopoeia (EP) Reference Standard