Merck
[The use of hydroxamic acids and sodium nitrate to enhance the antitumor effect of cyclophosphamide].
Voprosy onkologii (2013-07-03)
T N Bogatyrenko, Z V Kuropteva, T E Sashenkova, L M Baĭder, N P Konovalova
RESUMEN
It has been showed that the introduction of nitrocompounds (as nitic oxide donors) in to the compositions of cyclophosphamide and hydroxamic acids for curing animals having leukemia P-388 increased duration of life by 290%. Thereby 40% of animals have recovered. The therapeutic dose cyclophosphamide have been reduced by 6 times.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Cyclophosphamide monohydrate, bulk package
Sigma-Aldrich
Cyclophosphamide monohydrate, ISOPAC®
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium nitrate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%