Preparation, characterization and in vitro release study of gallic acid loaded silica nanoparticles for controlled release.
Die Pharmazie (2013-07-24)
Haifeng Hu, Lei Nie, Shuibin Feng, Jinping Suo
RESUMEN
Gallic acid (GA) is an interesting pharmaceutical component of plants. However, the short lifetime and the autoxidation of GA in aqueous solution significantly reduces its bioavailability and the residence time in the body system. In this study, GA was chemically bound to silica nanoparticles to control the release of GA based on the hydrolysis of the chemical bonds, and a silica nanoparticle drug delivery system was established. Gallic acid loaded silica nanoparticles (GA-SiO2) were synthesized by a modified Stober method. The Fourier Transform Infrared Spectroscopy (FTIR) and X-Ray Diffraction (XRD) analysis proved that GA did conjugate to silica nanoparticles. The particle size of the GA-SiO2 nanoparticles observed by Scanning Electron Microscope (SEM) was about 30 nm and the drug loading efficiency determined by Thermo Gravimetric Analysis/Differential Scanning Calorimetry (TGA/DSC) was 89.39%. The in vitro release study demonstrated that GA could be gradually released from the GA-SiO2. In addition, the antioxidant capability increased continuously during the immersion time, so the GA could serve as an excellent antioxidant to scavenge 1,1-diphenyl-2-picrylhydrazyl (DPPH) radicals in a long release period. Therefore, this study provided a novel drug delivery system for GA with controlled release capability and prolonged antioxidant activity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Supelco
Gallic acid, certified reference material, TraceCERT®
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
Silicon dioxide, alumina doped, nanoparticles, dispersion, <50 nm particle size, 20 wt. % in H2O, 99.99% trace metals basis