Merck
  • Inicio
  • Resultados de la búsqueda
  • Genotypic diversity and virulence traits of streptococcus sobrinus isolated from caries-free children and children suffering severe early childhood caries.
Genotypic diversity and virulence traits of streptococcus sobrinus isolated from caries-free children and children suffering severe early childhood caries.
The Chinese journal of dental research : the official journal of the Scientific Section of the Chinese Stomatological Association (CSA) (2013-07-24)
Xiu Rong Qin, Qiong Zhou, Man Qin
RESUMEN
To evaluate the genotypic diversity and some virulence traits of Streptococcus sobrinus (S. sobrinus) isolated from caries-free children and children suffering severe early childhood caries (SECC). S. sobrinus isolated from stimulated whole saliva samples of 91 caries-free children and 87 SECC children were subcultured, identified by polymerase chain reaction and genotyped by arbitrarily primed polymerase chain reaction. Polysaccharide synthesis ability, acidogenicity, aciduricity and the adherence ability of these S. sobrinus isolates were measured. The frequency of S. sobrinus detection was 18.39% (16/87) in SECC children, which was significantly higher than that (3.30%, 3/91) in caries-free children. One to three different genotypes of S. sobrinus were detected in each SECC child. Only one genotype was colonised in each caries-free child. In SECC children, the production of water-insoluble glucan (WIG) was positively correlated with the ability of S. sobrinus adhering to a glass surface. The presence of S. sobrinus could be a risk factor for high caries activity in severe early childhood caries. The multi-genotypes could be related to different caries suceptibility. Water-insoluble glucan plays an important role in the adherence and accumulation of S. sobrinus on tooth surfaces.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, analytical standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material