Merck
Metal solubilization from powdered printed circuit boards by microbial consortium from bauxite and pyrite ores.
Prikladnaia biokhimiia i mikrobiologiia (2013-07-26)
N N Adhapure, S S Waghmare, V S Hamde, A M Deshmukh
RESUMEN
With the current rapid developments in technology, there is an increasing accumulation of outdated electronic equipment. The primary reason for this increase is the low rate of recycling due to the complex nature of such waste. Bioleaching offers a promising solution for this problem. Study was conducted on the solubilization of heavy metals from electronic waste (e-waste). For this purpose, a microbial consortium from bauxite and pyrite ore samples was obtained using a simple "top down" approach. Essentially, printed circuit boards (PCB) were obtained and used as representative samples of e-waste. Various concentrations (1-5%) of PCB powder were subjected to bioleaching, and the effects on metal solubilization, changes in pH and concentration of ferrous iron produced were assessed. It was observed that a maximum of 96.93% Cu and 93.33% Zn was solubilized by microbial consortium from 10 g/l of PCB powder, whereas only 10.26% Ni was solubilized from 30 g/l of PCB powder. For lead, only 0.58% solubilization was achieved from 20 g/l of PCB powder. An analysis of the precipitate formed during bioleaching using scanning electron microscopy with energy dispersive x-ray analysis revealed the presence of Tin (59.96%), Cu (23.97%), Pb (9.30%) and Fe (5.92%).
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Iron, ≥99%, reduced, powder (fine)
Supelco
Aluminum oxide, for the determination of hydrocarbons
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Sigma-Aldrich
Carbonyl iron, ≥97% Fe basis
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Iron disulfide, powder, −325 mesh, 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, calcined, powder, primarily α-phase, 100-325 mesh
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm