Cystine plug and other novel mechanisms of large mechanical stability in dimeric proteins.
Physical review letters (2012-12-12)
Mateusz Sikora, Marek Cieplak
RESUMEN
We identify three dimeric proteins whose mechanostability is anisotropic and should exceed 1 nN along some directions. They come with distinct mechanical clamps: either shear-based, or involving a cystine slipknot, or due to dragging of a cystine plug through a cystine ring. The latter two mechanisms are topological in nature; the cystine plug mechanism has not yet been discussed but it turns out to provide the largest resistance to stretching. Its possible applications in elastomers are discussed.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
SAFC
L-Cystine
SAFC
L-Cystine
Supelco
L-Cystine, certified reference material, TraceCERT®
Cystine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%