Non-invasive study of human gall bladder bile in vivo using (1)H-MR spectroscopy.
The British journal of radiology (2003-07-15)
A S K Dzik-Jurasz, A P Prescot, M O Leach, D J Collins
RESUMEN
The sampling of gall bladder bile for analytical studies remains an invasive procedure. We demonstrate the application of the non-invasive methodology of (1)H-MR spectroscopy to the qualitative and quantitative assessment of human gall bladder bile in vivo. Spectral profiles in vivo are shown in relation to model and porcine gall bladder bile and the quantitation in man of the trimethylamine (choline) and lecithin concentrations were estimated to range from 25.9 mM to 48.4 mM (mean: 35.8 mM, standard deviation: 9.8). The composition of human gall bladder bile together with the quantitation of various constituents can be studied non-invasively in vivo.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Trimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in propylene glycol