Homogeneous enzymatic assay for L-cysteine with betaC-S lyase.
Bioscience, biotechnology, and biochemistry (2005-11-25)
Yoshiaki Nishiya, Yasuo Yoshida, Mamiko Yoshimura, Haruka Fukamachi, Yoshio Nakano
RESUMEN
We have developed a new enzymatic assay for determining L-cysteine concentration. The method involves the use of betaC-S lyase from Streptococcus anginosus, which catalyzes the alpha,beta-elimination of L-cysteine to hydrogen sulfide, pyruvate, and ammonia. The production of pyruvate is measured by D-lactate dehydrogenase and NADH. The decrease in NADH was proportional to the L-cysteine concentration up to 1.0 mM. When serum samples were used, within-day and day-to-day coefficient variations were below 4%. This method is simple, and can easily and reliably be used for accurate determination of L-cysteine concentration in serum or other samples.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Supelco
L-Cysteine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%