Merck
Folinic acid and enhanced renal elimination in formic acid intoxication.
Journal of toxicology. Clinical toxicology (1994-01-01)
D F Moore, A M Bentley, S Dawling, A M Hoare, J A Henry
RESUMEN
Ingestion of over 60 g of formic acid by an adult is potentially fatal. We report a case of a 36-year-old woman with a history of depression who ingested 110 g of formic acid. She survived a complicated intensive care hospitalization following usage of intravenous folinic acid, urinary alkalinization, intravenous furosemide and supportive care. We suggest a management protocol aimed at minimizing formate toxicity by enhancing hepatic formate degradation via the folinic acid 'one carbon pool' and by enhanced renal elimination of formate.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%