600 MHz 1H-NMR spectroscopy of human cerebrospinal fluid: effects of sample manipulation and assignment of resonances.
Journal of pharmaceutical and biomedical analysis (1993-08-01)
B C Sweatman, R D Farrant, E Holmes, F Y Ghauri, J K Nicholson, J C Lindon
RESUMEN
Extensive assignments of resonances in the 600 MHz 1H-NMR spectra of cerebrospinal fluid are reported. These have been achieved by the measurement of a combination of two-dimensional experiments comprising homonuclear J-resolved, COSY45, and double-quantum filtered COSY (DQCOSY) spectra. By these means the previous total of 18 endogenous metabolites, of which in general only selected resonances have been assigned, has been augmented to 46 molecules including all of the resonances of both alpha- and beta-anomers of glucose. With only a few exceptions all resonances have been assigned for all of the metabolites. In addition, the effect of freeze-drying on the 600 MHz 1H-NMR spectrum of human cerebrospinal fluid (CSF) is presented using both lyophilization with reconstitution into either H2O or D2O. Freeze-drying and reconstitution into H2O causes a significant sharpening of many small molecule resonances, including notably those of glutamate and glutamine as well as other amino acids and in addition causes the loss of volatile components, principally acetone. Further exchange of the H2O solvent by D2O causes no additional changes in the spectra.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
α-Ketoglutaric acid, 99.0-101.0% (T)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Dimethylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Oxaloacetic acid, ≥97% (HPLC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Dimethylamine solution, 2.0 M in THF
Sigma-Aldrich
Trimethylamine N-oxide, 95%
Sigma-Aldrich
Trimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
Oxaloacetic acid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97% (HPLC)
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Dimethylamine solution, 2.0 M in methanol
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%