• Inicio
  • Resultados de la búsqueda
  • Triplet-triplet energy transfer in fucoxanthin-chlorophyll protein from diatom Cyclotella meneghiniana: insights into the structure of the complex.
Triplet-triplet energy transfer in fucoxanthin-chlorophyll protein from diatom Cyclotella meneghiniana: insights into the structure of the complex.
Biochimica et biophysica acta (2013-07-17)
Marilena Di Valentin, Elena Meneghin, Laura Orian, Antonino Polimeno, Claudia Büchel, Enrico Salvadori, Christopher W M Kay, Donatella Carbonera
RESUMEN
Although the major light harvesting complexes of diatoms, called FCPs (fucoxanthin chlorophyll a/c binding proteins), are related to the cab proteins of higher plants, the structures of these light harvesting protein complexes are much less characterized. Here, a structural/functional model for the "core" of FCP, based on the sequence homology with LHCII, in which two fucoxanthins replace the central luteins and act as quenchers of the Chl a triplet states, is proposed. Combining the information obtained by time-resolved EPR spectroscopy on the triplet states populated under illumination, with quantum mechanical calculations, we discuss the chlorophyll triplet quenching in terms of the geometry of the chlorophyll-carotenoid pairs participating to the process. The results show that local structural rearrangements occur in FCP, with respect to LHCII, in the photoprotective site.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Chlorophyll a, from spinach
Sigma-Aldrich
Chlorophyll a, from Anacystis nidulans algae
Sigma-Aldrich
Fucoxanthin, carotenoid antioxidant