Merck
Enhanced glucose metabolism is preserved in cultured primary myotubes from obese donors in response to exercise training.
The Journal of clinical endocrinology and metabolism (2013-07-26)
Virginie Bourlier, Céline Saint-Laurent, Katie Louche, Pierre-Marie Badin, Claire Thalamas, Isabelle de Glisezinski, Dominique Langin, Coralie Sengenes, Cedric Moro
RESUMEN
It was suggested that human cultured primary myotubes retain the metabolic characteristics of their donor in vitro. The aim of the present study was to investigate whether the metabolic responses to endurance training are also conserved in culture. Middle-aged obese subjects completed an 8-week supervised aerobic exercise training program in which vastus lateralis muscle biopsies were collected before and after training. Anthropometric and blood parameters, as well as aerobic capacity, were assessed before and after training. Muscle biopsies were either used for Western blot analysis or digested to harvest myogenic progenitors that were differentiated into myotubes. Glucose oxidation, palmitate oxidation, and glycogen synthesis assays were performed on myotubes before and after training. Gene expression was assessed by real-time quantitative PCR. Our data indicate that in parallel of in vivo improvement of whole-body aerobic capacity and glucose metabolism, biopsy-derived primary myotubes showed similar patterns in vitro. Indeed, glucose oxidation, glycogen synthesis, and inhibition of palmitate oxidation by glucose were enhanced in myotubes after training. This was associated with consistent changes in the expression of metabolism-linked genes such as GLUT1, PDK4, and PDHA1. Interestingly, no difference in myogenic differentiation capacity was observed before and after training. Aerobic exercise training is associated with metabolic adaptations in vivo that are preserved in human cultured primary myotubes. It can be hypothesized that skeletal muscle microenvironmental changes induced by endurance training lead to metabolic imprinting on myogenic progenitor cells.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Glycogen from Mytilus edulis (Blue mussel), ≥85%
Sigma-Aldrich
Glycogen from Mytilus edulis (Blue mussel), for DNA precipitations
Sigma-Aldrich
Glycogen from rabbit liver, ≥85% dry basis (enzymatic)
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitic acid, ≥95%, FCC, FG
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Palmitic acid, certified reference material, TraceCERT®
Sigma-Aldrich
Pyruvate Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 0.1-1.0 units/mg protein (Lowry)
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Glycogen from bovine liver, ≥85%
Sigma-Aldrich
Glycogen from oyster, Type XI
Sigma-Aldrich
Glycogen from oyster, ≥75% dry basis