  • Search Results
  • Variations in genotype-phenotype correlations in phenylalanine hydroxylase deficiency in Chinese Han population.
Variations in genotype-phenotype correlations in phenylalanine hydroxylase deficiency in Chinese Han population.
Gene (2013-08-13)
Tianwen Zhu, Jun Ye, Lianshu Han, Wenjuan Qiu, Huiwen Zhang, Lili Liang, Xuefan Gu
ABSTRACT
The value of genotyping to predict variant phenotypes in patients with phenylalanine hydroxylase (Pah) deficiency is a matter of debate. However, there exists no comprehensive population relationship study focused on the Han Chinese. We analyzed genotype-phenotype correlation for 186 different genotypes in 338 unrelated Chinese patients harboring 109 different Pah mutations. Two systems were used in this process. The first was a phenotype prediction system based on arbitrary values (AV) attributed to each mutation. The second was a pair-wise correlation analysis. The observed phenotype for AV analysis was the corresponding metabolic phenotype stratified according to the pretreatment phenylalanine (Phe) value. We found that the observed phenotype matched the predicted phenotype in 54.41% of 272 patients for whom AV information was available; the highest degree of concordance (61.83%) was found in patients with null/null genotypes, whereas the lowest "concordance rate" (32.69%) was observed for patients with expected mild-PKU phenotype. There are repeated inconsistencies for such mutations as R241C, R243Q, R261Q, V388M, V399V, R408Q, A434D and EX6-96A>G which are associated with variable phenotypes in patients with identical genotype. Significant correlations were disclosed between pretreatment Phe values and predicted residual activity (r=-0.45643, P<0.0001) or AV sum (r=-0.59523, P<0.0001). Our study supports the notion that the Pah mutation genotype is the main determinant of metabolic phenotype in most patients in a particular population, and provided novel insights into the values that underpin the subsequent treatment and the prognosis of PKU in Chinese.
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Supelco
L-Phenylalanine, certified reference material, TraceCERT®
Supelco
L-Phenylalanine, analytical standard, for Nitrogen Determination According to Kjeldahl Method
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material