Derivatives of dibenzothiophene for positron emission tomography imaging of α7-nicotinic acetylcholine receptors.
Journal of medicinal chemistry (2013-09-21)
Yongjun Gao, Kenneth J Kellar, Robert P Yasuda, Thao Tran, Yingxian Xiao, Robert F Dannals, Andrew G Horti
RESUMEN
A new series of derivatives of 3-(1,4-diazabicyclo[3.2.2]nonan-4-yl)dibenzo[b,d]thiophene 5,5-dioxide with high binding affinities and selectivity for α7-nicotinic acetylcholine receptors (α7-nAChRs) (Ki = 0.4-20 nM) has been synthesized for positron emission tomography (PET) imaging of α7-nAChRs. Two radiolabeled members of the series [(18)F]7a (Ki = 0.4 nM) and [(18)F]7c (Ki = 1.3 nM) were synthesized. [(18)F]7a and [(18)F]7c readily entered the mouse brain and specifically labeled α7-nAChRs. The α7-nAChR selective ligand 1 (SSR180711) blocked the binding of [(18)F]7a in the mouse brain in a dose-dependent manner. The mouse blocking studies with non-α7-nAChR central nervous system drugs demonstrated that [(18)F]7a is highly α7-nAChR selective. In agreement with its binding affinity the binding potential of [(18)F]7a (BPND = 5.3-8.0) in control mice is superior to previous α7-nAChR PET radioligands. Thus, [(18)F]7a displays excellent imaging properties in mice and has been chosen for further evaluation as a potential PET radioligand for imaging of α7-nAChR in non-human primates.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
SAFC
L-Cystine
SAFC
L-Cystine
Supelco
L-Cystine, certified reference material, TraceCERT®
Cystine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%