  • Search Results
  • The effect of selected polymorphisms of the dopamine receptor gene DRD2 and the ANKK-1 on the preference of concentrations of sucrose solutions in men with alcohol dependence.
The effect of selected polymorphisms of the dopamine receptor gene DRD2 and the ANKK-1 on the preference of concentrations of sucrose solutions in men with alcohol dependence.
Psychiatria Danubina (2013-11-20)
Marcin Jabłoński, Andrzej Jasiewicz, Jolanta Kucharska-Mazur, Jerzy Samochowiec, Przemyslaw Bienkowski, Pawel Mierzejewski, Agnieszka Samochowiec
ABSTRACT
The aim of the study was to determine the influence of DRD2 gene polymorphisms in exon 8 G/A (rs 6276) in the promoter region -141 C Ins/Del (rs1799732) and the influence of ANKK-1 gene Taq-1A polymorphism (rs 1800497) on the preference of increasing sucrose concentrations in men with alcohol dependence. 63 male patients with alcohol dependence were genotyped for the above polymorphisms. Their preference for increasing sucrose concentrations was tested and their taste intensity perception of sucrose solutions was assessed. The patients were tested with the 'Sniffin' Sticks' olfactory test. We found a statistically significant association between some alleles of ANKK 1 gene Taq 1A polymorphisms and sucrose preference in the subjects. The A1 Taq 1A allele determined hedonistic response to the two highest concentrations of sucrose. No association was found regarding the other two polymorphisms (in the promoter region and in the exon 8 of the DRD2 gene). Study results suggest Taq-1A polymorphism plays a role in the preference to high concentrations of sucrose and its potential association with alcohol dependence pathogenesis.
Product Description
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material